Assay Details
| Gene Symbol: Rpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648850.1 | XP_007647040.1 | 285 | 55 | ||
| GenBank mRNA | JI880494.1 | - | 285 | 55 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpl28| Gene Symbol: Rpl28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648850.1 | XP_007647040.1 | 285 | 55 | ||
| GenBank mRNA | JI880494.1 | - | 285 | 55 | ||
Set Membership: |
|