Assay Details
Gene Symbol: Nifk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003498109.2 | XP_003498157.1 | 1 - 2 | 159 | 74 | |
GenBank mRNA | JI887407.1 | - | 1 - 2 | 152 | 74 | |
JP053254.1 | - | 1 - 2 | 152 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
