Assay Details
Gene Symbol: Gjd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003499534.1 | XP_003499582.1 | 1 - 2 | 66 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
