Assay Details
Gene Symbol: Caps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007643032.1 | XP_007641222.1 | 7 - 8 | 687 | 65 | |
XM_007643033.1 | XP_007641223.1 | 8 - 9 | 1037 | 65 | ||
XM_007643034.1 | XP_007641224.1 | 8 - 9 | 735 | 65 | ||
XM_007643035.1 | XP_007641225.1 | 7 - 8 | 682 | 65 | ||
XM_007643036.1 | XP_007641226.1 | 7 - 8 | 899 | 65 | ||
XM_007643037.1 | XP_007641227.1 | 6 - 7 | 553 | 65 | ||
XM_007643038.1 | XP_007641228.1 | 6 - 7 | 846 | 65 | ||
XM_007643040.1 | XP_007641230.1 | 8 - 9 | 740 | 65 | ||
XM_007643041.1 | XP_007641231.1 | 5 - 6 | 530 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
