Assay Details
Gene Symbol: Rita1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651392.1 | XP_007649582.1 | 2 - 3 | 949 | 82 | |
XM_007651393.1 | XP_007649583.1 | 2 - 3 | 949 | 82 | ||
XM_007651394.1 | XP_007649584.1 | 2 - 3 | 861 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
