Assay Details
Gene Symbol: Rita1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007651392.1 | XP_007649582.1 | 3 - 4 | 1482 | 62 | |
XM_007651394.1 | XP_007649584.1 | 3 - 4 | 1394 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() |
