Assay Details
| Gene Symbol: Gdnf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007644530.1 | XP_007642720.1 | 1 - 2 | 719 | 84 | |
| XM_007644531.1 | XP_007642721.1 | 1 - 2 | 719 | 84 | ||
More Information
Set Membership: |
|

Gdnf
Genomic Map