Assay Details
Gene Symbol: Slc39a11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495689.2 | XP_003495737.1 | 10 - 11 | 1063 | 60 | |
XM_003495690.2 | XP_003495738.1 | 10 - 11 | 1084 | 60 | ||
GenBank mRNA | JI886081.1 | - | 2 - 3 | 184 | 60 | |
JP062318.1 | - | 2 - 3 | 184 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
