Assay Details
Gene Symbol: Tcf21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512060.2 | XP_003512108.1 | 1 - 2 | 451 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
