Assay Details
Gene Symbol: Grwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003512191.2 | XP_003512239.1 | 1 - 2 | 376 | 69 | |
GenBank mRNA | JI865373.1 | - | 1 - 2 | 218 | 69 | |
JP045083.1 | - | 1 - 2 | 218 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
