Assay Details
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642830.1 | XP_007641020.1 | 2 - 3 | 247 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fabp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642830.1 | XP_007641020.1 | 2 - 3 | 247 | 86 |
Set Membership: |
![]() ![]() ![]() ![]() |