Assay Details
Gene Symbol: Stk35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007649679.1 | XP_007647869.1 | 1 - 2 | 1514 | 53 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
