Assay Details
| Gene Symbol: Npff | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507192.2 | XP_003507240.2 | 2 - 3 | 1811 | 105 | |
| XM_007647752.1 | XP_007645942.1 | 3 - 4 | 819 | 105 | ||
More Information
Set Membership: |
|

Npff
Genomic Map