Assay Details
Gene Symbol: Klhl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509663.2 | XP_003509711.1 | 2 - 3 | 1962 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Klhl25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003509663.2 | XP_003509711.1 | 2 - 3 | 1962 | 58 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |