Assay Details
| Gene Symbol: Mrpl30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003510866.2 | XP_003510914.1 | 2 - 3 | 192 | 94 | |
| GenBank mRNA | JI867002.1 | - | 3 - 4 | 267 | 94 | |
| JP065873.1 | - | 3 - 4 | 267 | 94 | ||
More Information
Set Membership: |
|

Mrpl30
Genomic Map