Assay Details
| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007624845.1 | XP_007623035.1 | 7 - 8 | 895 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Chrnb3| Gene Symbol: Chrnb3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007624845.1 | XP_007623035.1 | 7 - 8 | 895 | 64 | |
Set Membership: |
|