Assay Details
| Gene Symbol: Tmem194a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648082.1 | XP_007646272.1 | 4 - 5 | 529 | 105 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmem194a| Gene Symbol: Tmem194a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007648082.1 | XP_007646272.1 | 4 - 5 | 529 | 105 | |
Set Membership: |
|