Assay Details
Gene Symbol: Tmem194a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007648082.1 | XP_007646272.1 | 7 - 8 | 958 | 70 | |
GenBank mRNA | JI888812.1 | - | 3 - 4 | 433 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
