Assay Details
| Gene Symbol: Rab5a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007641215.1 | XP_007639405.1 | 2 - 3 | 924 | 71 | |
| XM_007641216.1 | XP_007639406.1 | 3 - 4 | 1020 | 71 | ||
| XM_007641217.1 | XP_007639407.1 | 4 - 5 | 1082 | 71 | ||
| XM_007641218.1 | XP_007639408.1 | 3 - 4 | 986 | 71 | ||
| GenBank mRNA | JI889307.1 | - | 2 - 3 | 338 | 71 | |
| JP060419.1 | - | 2 - 3 | 338 | 71 | ||
More Information
Set Membership: |
|

Rab5a
Genomic Map