Assay Details
Gene Symbol: Nkx3-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003496197.1 | XP_003496245.1 | 2 - 3 | 158 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
