Assay Details
| Gene Symbol: Bet1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496956.2 | XP_003497004.1 | 2 - 3 | 295 | 104 | |
| GenBank mRNA | JI867101.1 | - | 2 - 3 | 315 | 104 | |
| JP061341.1 | - | 2 - 3 | 315 | 104 | ||
More Information
Set Membership: |
|

Bet1
Genomic Map