Assay Details
Gene Symbol: Hint1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007640140.1 | XP_007638330.1 | 264 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
