Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003498639.2 | XP_003498687.2 | 6 - 7 | 1087 | 64 | |
| GenBank mRNA | JI891121.1 | - | 2 - 3 | 193 | 64 | |
More Information
Set Membership: |
|

Ciart
Genomic Map