Assay Details
| Gene Symbol: Srsf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003502018.2 | XP_003502066.2 | 2 - 3 | 361 | 71 | |
| GenBank mRNA | JI888138.1 | - | 2 - 3 | 337 | 71 | |
| JP059815.1 | - | 2 - 3 | 340 | 71 | ||
More Information
Set Membership: |
|

Srsf6
Genomic Map