Assay Details
| Gene Symbol: LOC100759306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504922.2 | XP_003504970.2 | 2 - 3 | 962 | 61 | |
| XM_007645415.1 | XP_007643605.1 | 2 - 3 | 175 | 61 | ||
| XM_007645416.1 | XP_007643606.1 | 2 - 3 | 383 | 61 | ||
More Information
Set Membership: |
|

LOC100759306
Genomic Map