Assay Details
| Gene Symbol: LOC100759306 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003504922.2 | XP_003504970.2 | 9 - 10 | 1722 | 85 | |
| XM_007645415.1 | XP_007643605.1 | 9 - 10 | 935 | 85 | ||
| XM_007645416.1 | XP_007643606.1 | 9 - 10 | 1143 | 85 | ||
More Information
Set Membership: |
|

LOC100759306
Genomic Map