Assay Details
Gene Symbol: Rsrc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003510295.2 | XP_003510343.1 | 6 - 7 | 525 | 80 | |
GenBank mRNA | JP055782.1 | - | 5 - 6 | 410 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
