Assay Details
Gene Symbol: Mrpl49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003514588.2 | XP_003514636.1 | 2 - 3 | 257 | 85 | |
GenBank mRNA | JI870243.1 | - | 2 - 3 | 234 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
