Assay Details
Gene Symbol: Acta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007653483.1 | XP_007651673.1 | 1 - 2 | 105 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
