Assay Details
Gene Symbol: Krtcap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007642246.1 | XP_007640436.1 | 4 - 5 | 326 | 62 | |
GenBank mRNA | JI876149.1 | - | 3 - 4 | 291 | 62 | |
JP048359.1 | - | 3 - 4 | 291 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
