Assay Details
Gene Symbol: Mrps28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007647957.1 | XP_007646147.1 | 2 - 3 | 198 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
