Assay Details
| Gene Symbol: Mrpl15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647066.1 | XP_007645256.1 | 2 - 3 | 172 | 83 | |
| GenBank mRNA | JI885421.1 | - | 1 - 2 | 62 | 83 | |
More Information
Set Membership: |
|

Mrpl15
Genomic Map