Assay Details
| Gene Symbol: Rbbp9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003496463.2 | XP_003496511.1 | 1 - 2 | 174 | 78 | |
| GenBank mRNA | JP063754.1 | - | 1 - 2 | 156 | 78 | |
More Information
Set Membership: |
|

Rbbp9
Genomic Map