Assay Details
Gene Symbol: Crebrf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003500821.2 | XP_003500869.1 | 4 - 5 | 1946 | 76 | |
XM_007641559.1 | XP_007639749.1 | 4 - 5 | 1946 | 76 | ||
XM_007641560.1 | XP_007639750.1 | 4 - 5 | 1946 | 76 | ||
XM_007641561.1 | XP_007639751.1 | 4 - 5 | 1946 | 76 | ||
XM_007641562.1 | XP_007639752.1 | 2 - 3 | 1285 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
