Assay Details
Gene Symbol: Krt78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654974.1 | XP_007653164.1 | 6 - 7 | 1849 | 60 | |
XM_007654975.1 | XP_007653165.1 | 7 - 8 | 1722 | 60 | ||
XM_007654976.1 | XP_007653166.1 | 5 - 6 | 1152 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
