Assay Details
Gene Symbol: LOC100774451 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007620022.1 | XP_007618212.1 | 2 - 3 | 737 | 99 | |
XM_007620030.1 | XP_007618220.1 | 2 - 3 | 468 | 99 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
