Assay Details
| Gene Symbol: LOC100774451 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007620022.1 | XP_007618212.1 | 4 - 5 | 2699 | 68 | |
| XM_007620030.1 | XP_007618220.1 | 4 - 5 | 2430 | 68 | ||
More Information
Set Membership: |
|

LOC100774451
Genomic Map