Assay Details
| Gene Symbol: Pet100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507745.2 | XP_003507793.1 | 2 - 3 | 148 | 67 | |
| XM_007648342.1 | XP_007646532.1 | 2 - 3 | 293 | 67 | ||
| XM_007648343.1 | XP_007646533.1 | 2 - 3 | 234 | 67 | ||
| XM_007648344.1 | XP_007646534.1 | 2 - 3 | 75 | 67 | ||
More Information
Set Membership: |
|

Pet100
Genomic Map