Assay Details
Gene Symbol: Hunk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003495576.2 | XP_003495624.1 | 4 - 5 | 583 | 62 | |
GenBank mRNA | JI876093.1 | - | 3 - 4 | 484 | 62 | |
JP054324.1 | - | 3 - 4 | 484 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
