Assay Details
Gene Symbol: Bloc1s5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003502006.2 | XP_003502054.1 | 3 - 4 | 342 | 76 | |
GenBank mRNA | JI865548.1 | - | 3 - 4 | 339 | 76 | |
JP055039.1 | - | 3 - 4 | 339 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
