Assay Details
| Gene Symbol: Rplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003509766.2 | XP_003509814.1 | 2 - 3 | 207 | 97 | |
| XM_007650422.1 | XP_007648612.1 | 2 - 3 | 207 | 97 | ||
| GenBank mRNA | JP040101.1 | - | 2 - 3 | 180 | 97 | |
More Information
Set Membership: |
|

Rplp2
Genomic Map