Assay Details
Gene Symbol: Rabepk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003511429.2 | XP_003511477.1 | 3 - 4 | 583 | 77 | |
XM_007651946.1 | XP_007650136.1 | 3 - 4 | 307 | 77 | ||
XM_007651947.1 | XP_007650137.1 | 3 - 4 | 547 | 77 | ||
XM_007651948.1 | XP_007650138.1 | 3 - 4 | 255 | 77 | ||
XM_007651949.1 | XP_007650139.1 | 3 - 4 | 586 | 77 | ||
XM_007651950.1 | XP_007650140.1 | 3 - 4 | 584 | 77 | ||
GenBank mRNA | JI880611.1 | - | 3 - 4 | 516 | 77 | |
JP037681.1 | - | 3 - 4 | 525 | 77 |

More Information
Set Membership: |
![]() ![]() ![]() |
