Assay Details
| Gene Symbol: Tnfsf12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007609214.1 | XP_007607404.1 | 5 - 6 | 355 | 58 | |
| GenBank mRNA | JP060443.1 | - | 1 - 2 | 91 | 58 | |
More Information
Set Membership: |
|

Tnfsf12
Genomic Map