Assay Details
Gene Symbol: Smug1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007654062.1 | XP_007652252.1 | 8 - 9 | 1455 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
