Assay Details
Gene Symbol: LOC100762989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007616046.1 | XP_007614236.1 | 3 - 4 | 544 | 105 | |
GenBank mRNA | JI867333.1 | - | 3 - 4 | 218 | 105 |

More Information
Set Membership: |
![]() ![]() |
