Assay Details
| Gene Symbol: LOC100767291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647332.1 | XP_007645522.1 | 2 - 3 | 1193 | 90 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100767291| Gene Symbol: LOC100767291 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007647332.1 | XP_007645522.1 | 2 - 3 | 1193 | 90 | |
Set Membership: |
|