Assay Details
Gene Symbol: Pink1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_007641585.1 | XP_007639775.1 | 1 - 2 | 333 | 75 | |
GenBank mRNA | JI893883.1 | - | 1 - 2 | 283 | 75 | |
JP043630.1 | - | 1 - 2 | 283 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
