Assay Details
| Gene Symbol: Lsm5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007645382.1 | XP_007643572.1 | 3 - 4 | 293 | 103 | |
| GenBank mRNA | JI896662.1 | - | 1 - 2 | 78 | 103 | |
More Information
Set Membership: |
|

Lsm5
Genomic Map