Assay Details
| Gene Symbol: Lhpp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003507285.2 | XP_003507333.1 | 3 - 4 | 521 | 63 | |
| GenBank mRNA | JI865411.1 | - | 1 - 2 | 115 | 63 | |
More Information
Set Membership: |
|

Lhpp
Genomic Map