Assay Details
| Gene Symbol: Gprc5b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_007653769.1 | XP_007651959.1 | 2 - 3 | 1184 | 74 | |
| XM_007653770.1 | XP_007651960.1 | 3 - 4 | 1109 | 74 | ||
More Information
Set Membership: |
|

Gprc5b
Genomic Map